Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199809.1 | NP_001186738.1 | 15 | 2210 | ||
NM_001199811.1 | NP_001186740.1 | 16 | 2357 | |||
NM_001199812.1 | NP_001186741.1 | 16 | 2357 | |||
NM_015434.3 | NP_056249.1 | 16 | 2357 | |||
NR_037667.1 | 17 | 2391 | ||||
XM_011509396.2 | XP_011507698.1 | 16 | 2242 | |||
GenBank mRNA | AK001363.1 | - | 16 | 2217 | ||
AK001598.1 | - | 16 | 2239 | |||
AK022509.1 | - | 16 | 2217 | |||
AK022589.1 | - | 8 | 1024 | |||
AK297225.1 | - | 15 | 2089 | |||
AL117576.1 | - | 5 | 631 | |||
AL133049.2 | - | 11 | 1526 | |||
BC020523.2 | - | 16 | 2233 | 3917030 | ||
BC030716.1 | - | 16 | 2212 | 3909231 | ||
BC033918.1 | - | 16 | 2216 | 3920829 |