Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098616.2 | NP_001092086.1 | 7 | 1008 | ||
NM_001297717.1 | NP_001284646.1 | 5 | 895 | |||
NM_001297718.1 | NP_001284647.1 | 7 | 931 | |||
NM_001297720.1 | NP_001284649.1 | 6 | 954 | |||
NM_015449.3 | NP_056264.1 | 6 | 906 | |||
NM_138740.3 | NP_620077.1 | 5 | 852 | |||
GenBank mRNA | AF077036.1 | - | 6 | 726 | ||
AF109185.1 | - | 7 | 699 | |||
AF529365.1 | - | 7 | 618 | |||
AJ243663.1 | - | 5 | 674 | |||
AJ243664.1 | - | 6 | 674 | |||
AJ243665.1 | - | 6 | 511 | |||
AL117463.1 | - | 5 | 472 | |||
BC000152.2 | - | 7 | 817 | 2901006 | ||
BC001377.2 | - | 6 | 674 | 3051048 | ||
BC006514.1 | - | 6 | 743 | 2822327 | ||
BC014391.1 | - | 6 | 642 | 3544590 | ||
BC088353.1 | - | 5 | 715 | 6164281 | ||
CR457131.1 | - | 6 | 516 | |||
DQ908899.1 | - | 6 | 516 |