Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NELFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015456.4 | NP_056271.3 | 13 | 2533 | ||
GenBank mRNA | AB033008.1 | - | 5 | 1215 | ||
AF464935.1 | - | 13 | 2338 | |||
AK022651.1 | - | 11 | 2029 | |||
AK091056.1 | - | 13 | 2526 | |||
AL050280.1 | - | 10 | 1780 | |||
BC011892.1 | - | 12 | 2242 | 3941719 | ||
BC019029.1 | - | 13 | 2492 |