Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NECAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015509.3 | NP_056324.2 | 5 | 559 | ||
NR_024260.1 | 4 | 477 | ||||
GenBank mRNA | AK074858.1 | - | 5 | 516 | ||
AK074880.1 | - | 5 | 529 | |||
AK074923.1 | - | 5 | 502 | |||
AK075013.1 | - | 5 | 536 | |||
AK094805.1 | - | 1 | 1014 | |||
AK290537.1 | - | 4 | 402 | |||
AK314424.1 | - | 4 | 411 | |||
AL050272.1 | - | 4 | 406 | |||
BC002888.2 | - | 5 | 461 | |||
BC067367.1 | - | 3 | 231 | |||
BC084551.1 | - | 1 | 70 | |||
BC110876.1 | - | 5 | 493 | 5553942 |