Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REXO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015523.3 | NP_056338.2 | 7 | 949 | ||
GenBank mRNA | AF151872.1 | - | 7 | 900 | ||
AK223200.1 | - | 7 | 864 | |||
AL110239.1 | - | 7 | 883 |