Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPATCH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015590.3 | NP_056405.2 | 2 | 125 | ||
NM_182679.2 | NP_872620.1 | 2 | 199 | |||
XM_005245287.3 | XP_005245344.1 | 2 | 208 | |||
GenBank mRNA | AK000256.1 | - | 2 | 192 | ||
AK125763.1 | - | 2 | 183 | |||
AK293026.1 | - | 2 | 193 | |||
AK301232.1 | - | 2 | 191 | |||
BC032436.1 | - | 2 | 178 | |||
BC040147.1 | - | 2 | 182 | |||
BF669837.1 | - | 194 | ||||
CB109133.1 | - | 2 | 108 | |||
DA087471.1 | - | 2 | 199 |