Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPATCH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015590.3 | NP_056405.2 | 5 | 347 | ||
NM_182679.2 | NP_872620.1 | 5 | 421 | |||
GenBank mRNA | AK000256.1 | - | 5 | 414 | ||
AK125763.1 | - | 4 | 331 | |||
AK293026.1 | - | 5 | 415 | |||
AK301232.1 | - | 5 | 413 | |||
BC032436.1 | - | 5 | 400 | |||
BC040147.1 | - | 5 | 404 | |||
BF669837.1 | - | 417 | ||||
CB109133.1 | - | 5 | 330 | |||
DA087471.1 | - | 5 | 421 |