Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278185.1 | NP_001265114.1 | 3 | 423 | ||
NM_001278186.1 | NP_001265115.1 | 3 | 423 | |||
NM_001278187.1 | NP_001265116.1 | 3 | 423 | |||
NM_015601.3 | NP_056416.2 | 3 | 423 | |||
NM_022079.2 | NP_071362.1 | 3 | 423 | |||
XM_011539592.2 | XP_011537894.1 | 3 | 425 | |||
XM_011539593.2 | XP_011537895.1 | 3 | 425 | |||
XM_011539594.2 | XP_011537896.1 | 3 | 378 | |||
XM_011539595.2 | XP_011537897.1 | 3 | 428 | |||
XM_011539596.2 | XP_011537898.1 | 3 | 378 | |||
XM_011539597.2 | XP_011537899.1 | 3 | 425 | |||
XM_017016041.1 | XP_016871530.1 | 3 | 425 | |||
GenBank mRNA | AK026808.1 | - | 3 | 360 | ||
AK292809.1 | - | 3 | 303 | |||
AY221963.1 | - | 3 | 360 | |||
AY650032.1 | - | 3 | 360 | |||
AY650033.1 | - | 3 | 360 | |||
AY650034.1 | - | 3 | 360 | |||
BC015043.1 | - | 3 | 326 | |||
BC039600.1 | - | 3 | 333 | 5212048 | ||
BX537563.1 | - | 1 | 101 | |||
DB459623.1 | - | 3 | 423 |