Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR1AIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267578.1 | NP_001254507.1 | 2 | 975 | ||
NM_015602.3 | NP_056417.2 | 2 | 975 | |||
XM_011509403.1 | XP_011507705.1 | 2 | 975 | |||
XM_011509404.1 | XP_011507706.1 | 2 | 975 | |||
GenBank mRNA | AK001780.1 | - | 2 | 975 | ||
AK021613.1 | - | 2 | 256 | |||
AK023204.1 | - | 2 | 206 | |||
AK291495.1 | - | 2 | 163 | |||
AK310012.1 | - | 2 | 321 | |||
BC023247.1 | - | 2 | 217 | 3458117 | ||
BQ435161.1 | - | 2 | 720 | |||
CN360425.1 | - | 2 | 189 |