Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SZRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114600.2 | NP_001108072.1 | 4 | 795 | ||
NM_001271869.1 | NP_001258798.1 | 4 | 792 | |||
NR_073500.1 | 2 | 490 | ||||
NR_073501.1 | 5 | 866 | ||||
NR_073502.1 | 3 | 745 | ||||
NR_073503.1 | 3 | 742 | ||||
GenBank mRNA | AB097042.1 | - | 3 | 533 | ||
AB097043.1 | - | 3 | 533 | |||
AB097044.1 | - | 3 | 524 | |||
AB097045.1 | - | 3 | 524 | |||
AB097046.1 | - | 4 | 583 | |||
AJ295987.1 | - | 3 | 529 | |||
AK297926.1 | - | 5 | 547 | |||
AK298576.1 | - | 4 | 479 | |||
AK308787.1 | - | 2 | 277 | |||
AK316205.1 | - | 4 | 469 | |||
AL050028.1 | - | 3 | 529 | |||
BC023988.1 | - | 3 | 550 | 3825796 | ||
BC065538.1 | - | 3 | 530 | 6053254 |