Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002901.2 | NP_002892.1 | 1, 2 | 517 | ||
GenBank mRNA | AK129791.1 | - | 1, 2 | 544 | ||
BC010120.2 | - | 1, 2 | 386 | 3836263 | ||
BP253627.1 | - | 1, 2 | 517 | |||
D42073.1 | - | 1, 2 | 303 | |||
FJ224346.1 | - | 1, 2 | 303 |