Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCAF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015726.3 | NP_056541.2 | 10 | 1507 | ||
NR_028103.1 | 10 | 1807 | ||||
NR_028104.1 | 9 | 1733 | ||||
GenBank mRNA | AK026481.1 | - | 10 | 1573 | ||
AK223475.1 | - | 9 | 1362 | |||
AK303188.1 | - | 11 | 1375 | |||
AK312690.1 | - | 9 | 1656 | |||
BC098271.1 | - | 10 | 1413 | 40001575 | ||
BC099709.1 | - | 10 | 1413 | 40001574 | ||
BC099846.1 | - | 10 | 1413 | 40001581 | ||
CR749801.1 | - | 10 | 1730 | |||
U06631.1 | - | 10 | 1471 |