Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCAF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015726.3 | NP_056541.2 | 14 | 2932 | ||
NR_028103.1 | 14 | 3232 | ||||
NR_028104.1 | 13 | 3158 | ||||
GenBank mRNA | AK026481.1 | - | 16 | 2427 | ||
U06631.1 | - | 14 | 2885 |