Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270764.1 | NP_001257693.1 | 8 | 3693 | ||
NM_015892.4 | NP_056976.2 | 8 | 3655 | |||
XM_005269891.3 | XP_005269948.1 | 8 | 3771 | |||
XM_006717891.3 | XP_006717954.1 | 8 | 4088 | |||
XM_017016319.1 | XP_016871808.1 | 8 | 3607 | |||
GenBank mRNA | AB011170.1 | - | 8 | 3623 | ||
AF026477.1 | - | 7 | 3249 | |||
AK299546.1 | - | 4 | 1449 | |||
BC027908.1 | - | 8 | 3598 | 5204516 | ||
BC075813.1 | - | 7 | 3351 | 5213653 | ||
CR749804.1 | - | 8 | 3776 |