Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007239.1 | NP_001007240.1 | 8 | 2858 | ||
NM_014955.2 | NP_055770.1 | 8 | 3001 | |||
NM_015935.4 | NP_057019.3 | 8 | 3326 | |||
XM_017001457.1 | XP_016856946.1 | 6 | 2601 | |||
XM_017001458.1 | XP_016856947.1 | 7 | 3278 | |||
XM_017001459.1 | XP_016856948.1 | 7 | 2828 | |||
GenBank mRNA | AB242175.1 | - | 8 | 3239 | ||
AK027621.1 | - | 8 | 2940 | |||
AK094818.1 | - | 1 | 2365 | |||
AK129561.1 | - | 7 | 2674 | |||
AL049669.1 | - | 8 | 2783 | |||
BX094025.1 | - | 1 | 646 |