Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323625.1 | NP_001310554.1 | 9 | 1628 | ||
NM_016361.4 | NP_057445.4 | 9 | 1597 | |||
NR_136633.1 | 9 | 1583 | ||||
NR_136634.1 | 10 | 1958 | ||||
NR_136635.1 | 9 | 1583 | ||||
NR_136636.1 | 9 | 1903 | ||||
XM_011509601.2 | XP_011507903.1 | 9 | 1553 | |||
XM_017001422.1 | XP_016856911.1 | 9 | 1553 | |||
GenBank mRNA | AB030039.2 | - | 9 | 1401 | ||
AB031478.1 | - | 9 | 1508 | |||
AB209248.1 | - | 8 | 3679 | |||
AK000657.1 | - | 9 | 1334 | |||
AY358552.1 | - | 9 | 1305 | |||
BC009965.1 | - | 9 | 1577 | 4299943 | ||
CR457101.1 | - | 9 | 1112 | |||
KJ534759.1 | - | 9 | 1133 | |||
KJ535036.1 | - | 8 | 1005 | |||
KJ535044.1 | - | 6 | 771 | |||
KJ535055.1 | - | 9 | 1088 |