Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIKESHI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322404.1 | NP_001309333.1 | 3 | 525 | ||
NM_001322407.1 | NP_001309336.1 | 4 | 699 | |||
NM_001322409.1 | NP_001309338.1 | 4 | 629 | |||
NM_016401.3 | NP_057485.2 | 3 | 541 | |||
NR_024597.1 | 2 | 303 | ||||
NR_024598.1 | 2 | 303 | ||||
NR_136324.1 | 4 | 649 | ||||
XM_017017914.1 | XP_016873403.1 | 3 | 531 | |||
XM_017017915.1 | XP_016873404.1 | 3 | 318 | |||
GenBank mRNA | AA148558.1 | - | 2 | 268 | ||
AF151082.1 | - | 2 | 184 | |||
AF161487.1 | - | 3 | 522 | |||
AF161527.1 | - | 3 | 525 | |||
AK001447.1 | - | 3 | 512 | |||
AK223198.1 | - | 3 | 512 | |||
AK298763.1 | - | 4 | 665 | |||
BC001677.2 | - | 3 | 414 | 2820772 | ||
BC006476.2 | - | 3 | 414 | 2820772 | ||
BC015991.2 | - | 3 | 506 | 2820691 | ||
BC018080.1 | - | 3 | 475 | 4798958 | ||
BC021621.1 | - | 3 | 506 | 2820691 | ||
BP423222.1 | - | 2 | 220 | |||
BX332668.2 | - | 3 | 378 |