Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286229.1 | NP_001273158.1 | 4 | 605 | ||
NM_001286230.1 | NP_001273159.1 | 4 | 605 | |||
NM_016448.3 | NP_057532.3 | 5 | 731 | |||
XM_011509614.1 | XP_011507916.1 | 5 | 435 | |||
GenBank mRNA | AF195765.1 | - | 5 | 495 | ||
AF345896.1 | - | 5 | 516 | |||
AK000742.1 | - | 5 | 549 | |||
AK027651.1 | - | 4 | 312 | |||
AK292343.1 | - | 5 | 603 | |||
AK303346.1 | - | 4 | 408 | |||
BC033297.1 | - | 5 | 472 | 4342989 | ||
BC033540.1 | - | 5 | 513 | 4826434 | ||
DQ641253.1 | - | 5 | 393 |