Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099431.1 | NP_001092901.1 | 4 | 560 | ||
NM_016509.3 | NP_057593.3 | 5 | 659 | |||
XM_005253382.4 | XP_005253439.1 | 7 | 1031 | |||
XM_011520685.2 | XP_011518987.1 | 6 | 940 | |||
XM_011520686.2 | XP_011518988.1 | 5 | 568 | |||
XM_017019395.1 | XP_016874884.1 | 6 | 1015 | |||
XM_017019396.1 | XP_016874885.1 | 4 | 367 | |||
GenBank mRNA | AF124841.1 | - | 5 | 516 | ||
AY358599.1 | - | 5 | 640 | |||
BC029554.1 | - | 6 | 631 | 5269252 | ||
BX647321.1 | - | 4 | 351 |