Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297748.1 | NP_001284677.1 | 5 | 1056 | ||
NM_001297749.1 | NP_001284678.1 | 4 | 879 | |||
NM_001297750.1 | NP_001284679.1 | 7 | 1253 | |||
NM_001297751.1 | NP_001284680.1 | 6 | 1197 | |||
NM_016511.3 | NP_057595.2 | 6 | 1155 | |||
XM_011520687.2 | XP_011518989.1 | 7 | 1236 | |||
GenBank mRNA | AK075114.1 | - | 6 | 1075 | ||
AK091989.1 | - | 5 | 1007 | |||
AK300979.1 | - | 4 | 766 | |||
AY358587.1 | - | 6 | 1155 | |||
BC039072.1 | - | 6 | 1061 | 5178017 | ||
BC067746.1 | - | 6 | 1075 | 30337675 |