Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NTM SIRNA Assays ›
Gene Symbol:
NTM
Gene Name:
neurotrimin
Gene Aliases:
HNT, IGLON2, NTRI
Chromosome Location:
Chr.11: 131370326 - 132336822 on Build GRCh38
UniGene ID:
Hs.504352
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
122929
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NTM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001048209.1
NP_001041674.1
3
635
NM_001144058.1
NP_001137530.1
2
968
NM_001144059.1
NP_001137531.1
2
968
NM_016522.2
NP_057606.1
2
968
XM_005271580.3
XP_005271637.1
3
785
XM_005271581.3
XP_005271638.1
2
738
XM_005271582.3
XP_005271639.1
3
785
XM_006718855.3
XP_006718918.1
4
892
XM_006718857.3
XP_006718920.1
3
812
XM_011542861.2
XP_011541163.1
2
738
XM_017017851.1
XP_016873340.1
2
738
XM_017017852.1
XP_016873341.1
3
785
XM_017017853.1
XP_016873342.1
4
892
XM_017017854.1
XP_016873343.1
2
738
XM_017017855.1
XP_016873344.1
3
785
XM_017017856.1
XP_016873345.1
4
498
XM_017017857.1
XP_016873346.1
4
789
XM_017017858.1
XP_016873347.1
4
782
XM_017017859.1
XP_016873348.1
4
498
XM_017017860.1
XP_016873349.1
3
815
XM_017017861.1
XP_016873350.1
4
892
XM_017017862.1
XP_016873351.1
4
779
XM_017017863.1
XP_016873352.1
3
815
XM_017017864.1
XP_016873353.1
3
783
XM_017017865.1
XP_016873354.1
3
785
XM_017017866.1
XP_016873355.1
3
812
GenBank mRNA
AF126426.1
-
2
568
AK293448.1
-
3
603
AK293487.1
-
4
584
AK294137.1
-
3
514
AK315964.1
-
3
630
AY358331.1
-
3
437
BC050716.1
-
2
318
6166839
BE263639.1
-
1
34
EF043567.1
-
2
324
Back To Top
More Information
Panther Classification:
Molecular Function -
immunoglobulin
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
neuron recognition
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-9bfc7:80/100.66.79.6:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline