Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERGIC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016570.2 | NP_057654.2 | 10 | 813 | ||
GenBank mRNA | AF183410.1 | - | 10 | 767 | ||
AF216751.1 | - | 10 | 771 | |||
AF302767.1 | - | 10 | 766 | |||
AK074520.1 | - | 10 | 788 | |||
AK222799.1 | - | 10 | 753 | |||
AL834128.2 | - | 10 | 754 | |||
BC000887.1 | - | 10 | 766 | 3462408 | ||
BC107794.1 | - | 10 | 770 | 30319807 |