Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERGIC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016570.2 | NP_057654.2 | 11, 12 | 965 | ||
GenBank mRNA | AF183410.1 | - | 11, 12 | 919 | ||
AF216751.1 | - | 11, 12 | 923 | |||
AF302767.1 | - | 11, 12 | 918 | |||
AK074520.1 | - | 11, 12 | 940 | |||
AK222799.1 | - | 11, 12 | 905 | |||
AL834128.2 | - | 11, 12 | 906 | |||
BC000887.1 | - | 11, 12 | 918 | 3462408 | ||
BC107794.1 | - | 11, 12 | 922 | 30319807 |