Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002922.3 | NP_002913.3 | 2, 3 | 287 | ||
GenBank mRNA | AF493925.1 | - | 2, 3 | 182 | ||
AK093544.1 | - | 2, 3 | 239 | |||
AK303138.1 | - | 2, 3 | 241 | |||
AK315607.1 | - | 2, 3 | 241 | |||
AL039059.1 | - | 2, 3 | 291 | |||
BC015510.1 | - | 2, 3 | 219 | 3916789 | ||
BG536972.1 | - | 2, 3 | 287 | |||
BT006668.1 | - | 2, 3 | 182 | |||
EU668340.1 | - | 2, 3 | 219 | |||
X73427.1 | - | 2, 3 | 196 |