Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127350.1 | NP_001120822.1 | 3 | 356 | ||
NM_017638.2 | NP_060108.2 | 3 | 412 | |||
XM_005245914.4 | XP_005245971.1 | 3 | 326 | |||
GenBank mRNA | AB107222.1 | - | 3 | 368 | ||
AK000052.1 | - | 3 | 376 | |||
AK296130.1 | - | 3 | 403 | |||
BC002694.2 | - | 3 | 359 | 3607295 | ||
BG752399.1 | - | 3 | 357 | |||
DA090416.1 | - | 3 | 444 |