Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017824.4 | NP_060294.1 | 3, 4 | 709 | ||
XM_011539886.2 | XP_011538188.1 | 4, 5 | 857 | |||
GenBank mRNA | AB191202.1 | - | 3, 4 | 465 | ||
AK000452.1 | - | 3, 4 | 711 | |||
AK294799.1 | - | 3, 4 | 783 | |||
BC015480.1 | - | 3, 4 | 669 | 3905766 |