Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164245.1 | NP_001157717.1 | 10 | 1095 | ||
NM_001164246.1 | NP_001157718.1 | 10 | 1068 | |||
NM_017847.5 | NP_060317.3 | 11 | 1164 | |||
XM_011509660.1 | XP_011507962.1 | 11 | 1163 | |||
XM_011509661.1 | XP_011507963.1 | 11 | 1163 | |||
GenBank mRNA | AK000512.1 | - | 11 | 1084 | ||
AK026086.1 | - | 9 | 734 | |||
AK298107.1 | - | 10 | 1004 | |||
AY854248.1 | - | 11 | 1057 | |||
BC003397.1 | - | 11 | 997 | 3448518 | ||
DQ323046.1 | - | 10 | 928 | |||
DQ529299.1 | - | 9 | 859 |