Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317338.1 | NP_001304267.1 | 5, 6 | 1106 | ||
NM_017898.4 | NP_060368.2 | 5, 6 | 1106 | |||
XM_006711407.2 | XP_006711470.1 | 4, 5 | 1662 | |||
XM_011509684.1 | XP_011507986.1 | 5, 6 | 630 | |||
GenBank mRNA | AK000612.1 | - | 3, 4 | 245 | ||
BC010366.1 | - | 4, 5 | 440 | |||
BC011973.1 | - | 5, 6 | 1018 | 3458649 | ||
BC015829.2 | - | 3, 4 | 376 | |||
BC016859.1 | - | 5, 6 | 1008 | 3849257 |