Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNHIT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170670.1 | NP_001164141.1 | 11 | 2344 | ||
NM_017953.3 | NP_060423.3 | 10 | 2461 | |||
GenBank mRNA | AF308296.1 | - | 10 | 2108 | ||
AK000736.1 | - | 10 | 1833 | |||
AK000767.1 | - | 10 | 2370 | |||
AL442074.1 | - | 10 | 2446 | |||
BC026236.1 | - | 10 | 2449 | 4664103 | ||
BC110898.1 | - | 10 | 2378 | 6012059 |