Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PACS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018026.3 | NP_060496.2 | 3 | 613 | ||
XM_011545162.1 | XP_011543464.1 | 3 | 173 | |||
XM_011545164.2 | XP_011543466.1 | 3 | 1481 | |||
GenBank mRNA | AK056361.1 | - | 2 | 120 | ||
AK096644.1 | - | 3 | 613 | |||
AK098336.1 | - | 4 | 582 | |||
AY320283.1 | - | 3 | 653 | |||
BC010096.1 | - | 3 | 521 | 3502170 | ||
BC052577.1 | - | 3 | 512 | 4130626 |