Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PACS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018026.3 | NP_060496.2 | 13, 14 | 1764 | ||
XM_011545162.1 | XP_011543464.1 | 13, 14 | 1324 | |||
XM_011545164.2 | XP_011543466.1 | 13, 14 | 2632 | |||
GenBank mRNA | AB033001.1 | - | 6, 7 | 666 | ||
AK001071.1 | - | 1, 2 | 69 | |||
AK056361.1 | - | 12, 13 | 1271 | |||
AK096644.1 | - | 12, 13 | 1614 | |||
AK293966.1 | - | 2, 3 | 375 | |||
AY320283.1 | - | 13, 14 | 1804 | |||
BC010096.1 | - | 13, 14 | 1672 | 3502170 | ||
BC042909.1 | - | 2, 3 | 223 | |||
BC052577.1 | - | 13, 14 | 1663 | 4130626 | ||
BC055288.1 | - | 5, 6 | 468 |