Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300958.1 | NP_001287887.1 | 10 | 2092 | ||
XM_011541677.2 | XP_011539979.1 | 10 | 2339 | |||
GenBank mRNA | BX641056.1 | - | 9 | 2058 | ||
DA573011.1 | - | 1 | 361 |