Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XKR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018053.3 | NP_060523.2 | 3 | 1740 | ||
XM_011541679.2 | XP_011539981.1 | 5 | 2011 | |||
XM_011541680.2 | XP_011539982.1 | 4 | 1903 | |||
GenBank mRNA | AK001169.1 | - | 1 | 407 | ||
AK026024.1 | - | 3 | 1209 | |||
AK054909.1 | - | 1 | 1833 | |||
AK091615.1 | - | 3 | 1740 | |||
AY534246.1 | - | 3 | 1198 | |||
BC003606.1 | - | 1 | 627 | |||
BC013379.2 | - | 3 | 1199 | 4025274 | ||
BC028564.2 | - | 3 | 1199 | 3832072 |