Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168551.1 | NP_001162023.1 | 14 | 1785 | ||
NM_018087.4 | NP_060557.3 | 14 | 1905 | |||
NR_033142.1 | 13 | 1784 | ||||
XM_011541766.2 | XP_011540068.1 | 14 | 1747 | |||
GenBank mRNA | AK001269.1 | - | 14 | 1788 | ||
AK022618.1 | - | 2 | 61 | |||
AK091439.1 | - | 8 | 591 | |||
AK295000.1 | - | 14 | 1569 | |||
AK298909.1 | - | 13 | 1528 | |||
AK302910.1 | - | 12 | 1363 | |||
AL354613.1 | - | 14 | 1785 | |||
BC003082.2 | - | 14 | 1855 | 3509727 | ||
BX648031.1 | - | 13 | 1515 | |||
DQ141696.1 | - | 14 | 1614 |