Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NECAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145277.1 | NP_001138749.1 | 7 | 1919 | ||
NM_001145278.1 | NP_001138750.1 | 8 | 1917 | |||
NM_018090.4 | NP_060560.1 | 8 | 1995 | |||
XM_011541767.2 | XP_011540069.1 | 8 | 1991 | |||
XM_017001721.1 | XP_016857210.1 | 7 | 1788 | |||
GenBank mRNA | AK001282.1 | - | 9 | 1716 | ||
AK021938.1 | - | 4 | 1471 | |||
AK023545.1 | - | 7 | 1855 | |||
AK024468.1 | - | 7 | 4547 | |||
AK223344.1 | - | 9 | 1217 | |||
BC017014.1 | - | 8 | 1926 | 3848563 | ||
BC018914.2 | - | 8 | 1929 | 4299890 |