Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307977.1 | NP_001294906.1 | 7, 8 | 812 | ||
NM_018093.3 | NP_060563.2 | 8, 9 | 891 | |||
XM_005274055.2 | XP_005274112.1 | 8, 9 | 2752 | |||
GenBank mRNA | AK001301.1 | - | 8, 9 | 891 | ||
AK025383.1 | - | 3, 4 | 326 | |||
AK292330.1 | - | 8, 9 | 1261 | |||
AY500828.1 | - | 7, 8 | 743 | |||
BC006351.2 | - | 7, 8 | 734 | 4111690 |