Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320047.1 | NP_001306976.1 | 6 | 588 | ||
NM_001320048.1 | NP_001306977.1 | 4 | 380 | |||
NM_001320050.1 | NP_001306979.1 | 5 | 469 | |||
NM_001320051.1 | NP_001306980.1 | 5 | 492 | |||
NM_018186.3 | NP_060656.2 | 5 | 891 | |||
XM_005245317.4 | XP_005245374.1 | 5 | 463 | |||
XM_011509735.2 | XP_011508037.1 | 3 | 212 | |||
XM_017001722.1 | XP_016857211.1 | 5 | 463 | |||
XM_017001723.1 | XP_016857212.1 | 5 | 463 | |||
XM_017001724.1 | XP_016857213.1 | 3 | 212 | |||
XM_017001725.1 | XP_016857214.1 | 3 | 212 | |||
GenBank mRNA | AK001568.1 | - | 4 | 300 | ||
AK096493.1 | - | 6 | 498 | |||
AK127098.1 | - | 9 | 1322 | |||
AK294569.1 | - | 1 | 34 | |||
AK299369.1 | - | 5 | 374 | |||
AK303296.1 | - | 4 | 339 | |||
AK310638.1 | - | 4 | 289 | |||
AL354614.1 | - | 5 | 891 | |||
BC062689.1 | - | 3 | 182 | |||
BC091516.1 | - | 6 | 502 | 30389801 | ||
BC107168.1 | - | 5 | 379 | 40035373 | ||
BC107169.1 | - | 5 | 379 | 40035375 | ||
CB159947.1 | - | 2 | 111 | |||
DC401980.1 | - | 5 | 470 |