Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018198.3 | NP_060668.2 | 13 | 1452 | ||
XM_017001745.1 | XP_016857234.1 | 12 | 1305 | |||
GenBank mRNA | AF306695.1 | - | 13 | 1366 | ||
AK001599.1 | - | 13 | 1361 | |||
AK025491.1 | - | 3 | 126 | |||
AK093709.1 | - | 1 | 607 | |||
AK095386.1 | - | 4 | 1174 | |||
AK294539.1 | - | 12 | 1275 | |||
AK298378.1 | - | 13 | 1338 | |||
AL833841.1 | - | 13 | 1326 | |||
AY336749.1 | - | 13 | 1362 | |||
BC006086.1 | - | 13 | 1348 | 3946414 | ||
BC008772.1 | - | 13 | 1353 | |||
BC014145.2 | - | 12 | 1204 | 4762069 |