Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018198.3 | NP_060668.2 | 5 | 525 | ||
XM_017001745.1 | XP_016857234.1 | 4 | 378 | |||
GenBank mRNA | AF306694.1 | - | 5 | 424 | ||
AF306695.1 | - | 5 | 439 | |||
AF306696.1 | - | 3 | 229 | |||
AK001599.1 | - | 5 | 434 | |||
AK294539.1 | - | 4 | 348 | |||
AK298378.1 | - | 5 | 411 | |||
AL109978.1 | - | 4 | 347 | |||
AL833841.1 | - | 5 | 399 | |||
AY336749.1 | - | 5 | 435 | |||
BC006086.1 | - | 5 | 421 | 3946414 | ||
BC008772.1 | - | 5 | 426 | |||
BC014145.2 | - | 5 | 433 | 4762069 |