Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253379.1 | NP_001240308.1 | 3 | 632 | ||
NM_001253380.1 | NP_001240309.1 | 2 | 394 | |||
NM_001253382.1 | NP_001240311.1 | 3 | 383 | |||
NM_001253383.1 | NP_001240312.1 | 3 | 362 | |||
NM_001253384.1 | NP_001240313.1 | 3 | 619 | |||
NM_002948.3 | NP_002939.2 | 3 | 619 | |||
GenBank mRNA | AF279903.1 | - | 2 | 273 | ||
AF283772.2 | - | 3 | 279 | |||
AK091751.1 | - | 1 | 1359 | |||
AK098795.1 | - | 3 | 269 | |||
AK129815.1 | - | 3 | 279 | |||
AK290694.1 | - | 3 | 275 | |||
AK292000.1 | - | 3 | 275 | |||
AK293709.1 | - | 3 | 275 | |||
AK297091.1 | - | 3 | 277 | |||
AY347528.1 | - | 2 | 344 | |||
BC014837.1 | - | 3 | 282 | 2901238 | ||
BC068198.1 | - | 1 | 278 | 4093343 | ||
BC070328.1 | - | 3 | 275 | 30371842 | ||
BC071672.1 | - | 1 | 264 | 4390593 | ||
BC081565.1 | - | 3 | 284 | 5458912 | ||
BE893727.1 | - | 3 | 354 | |||
BT019672.1 | - | 2 | 239 | |||
BU178524.1 | - | 3 | 383 | |||
BX537495.1 | - | 2 | 683 | |||
DA037869.1 | - | 3 | 300 | |||
DA205054.1 | - | 3 | 345 | |||
DB461802.1 | - | 2 | 394 | |||
DC351786.1 | - | 362 | ||||
L25899.1 | - | 2 | 255 |