Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002949.3 | NP_002940.2 | 4 | 581 | ||
GenBank mRNA | AF105278.1 | - | 3 | 314 | ||
BC002344.2 | - | 4 | 566 | 2961288 | ||
BC007497.2 | - | 4 | 463 | 2959986 | ||
U25041.1 | - | 4 | 467 | |||
X79865.1 | - | 4 | 586 |