Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSBN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018364.4 | NP_060834.2 | 7 | 2728 | ||
NR_130896.1 | 8 | 2846 | ||||
GenBank mRNA | AK022166.1 | - | 6 | 1907 | ||
BC026155.1 | - | 8 | 2668 | 4827655 | ||
CR627402.1 | - | 8 | 2321 |