Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STYK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018423.2 | NP_060893.2 | 11 | 2327 | ||
XM_005253417.2 | XP_005253474.1 | 10 | 2227 | |||
XM_011520736.1 | XP_011519038.1 | 12 | 2130 | |||
XM_011520737.2 | XP_011519039.1 | 11 | 2167 | |||
XM_011520738.2 | XP_011519040.1 | 11 | 2322 | |||
XM_017019579.1 | XP_016875068.1 | 11 | 2110 | |||
GenBank mRNA | AF251059.1 | - | 11 | 2327 | ||
AL353940.1 | - | 11 | 2075 |