Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCSER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284240.1 | NP_001271169.1 | 3 | 1750 | ||
NM_001284241.1 | NP_001271170.1 | 3 | 1750 | |||
NM_018999.3 | NP_061872.2 | 3 | 1750 | |||
XM_005269905.3 | XP_005269962.1 | 3 | 1781 | |||
XM_005269906.3 | XP_005269963.1 | 3 | 1781 | |||
XM_011539870.2 | XP_011538172.1 | 3 | 1781 | |||
XM_011539871.2 | XP_011538173.1 | 3 | 1781 | |||
XM_011539872.2 | XP_011538174.1 | 3 | 1766 | |||
XM_017016340.1 | XP_016871829.1 | 3 | 1670 | |||
XM_017016341.1 | XP_016871830.1 | 3 | 1766 | |||
XM_017016342.1 | XP_016871831.1 | 3 | 1748 | |||
XM_017016343.1 | XP_016871832.1 | 2 | 1511 | |||
GenBank mRNA | AB032954.1 | - | 2 | 753 | ||
AF241785.1 | - | 2 | 1239 | |||
AK024324.1 | - | 2 | 1474 | |||
AK298966.1 | - | 3 | 1676 | |||
BC030528.1 | - | 3 | 1648 | 5208702 | ||
CR991891.1 | - | 2 | 403 |