Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L1TD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164835.1 | NP_001158307.1 | 4 | 929 | ||
NM_019079.4 | NP_061952.3 | 3 | 818 | |||
GenBank mRNA | AB211065.1 | - | 3 | 756 | ||
AK001746.1 | - | 1 | 187 | |||
AK001973.1 | - | 3 | 744 | |||
AL834314.1 | - | 3 | 756 | |||
BC111559.1 | - | 3 | 731 | 40035707 | ||
CX167172.1 | - | 3 | 819 |