Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002961.2 | NP_002952.1 | 3 | 223 | ||
NM_019554.2 | NP_062427.1 | 4 | 272 | |||
GenBank mRNA | AK292083.1 | - | 3 | 223 | ||
AY762982.1 | - | 126 | ||||
BC000838.1 | - | 4 | 256 | 3456833 | ||
BC016300.1 | - | 3 | 227 | 4247807 | ||
BC024974.1 | - | 3 | 207 | |||
CB109749.1 | - | 4 | 272 | |||
CR450345.1 | - | 2 | 154 | |||
M80563.1 | - | 3 | 289 |