Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRTAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019604.3 | NP_062550.2 | 5 | 635 | ||
XM_011542900.2 | XP_011541202.1 | 4 | 561 | |||
GenBank mRNA | AF001622.1 | - | 5 | 586 | ||
AK313578.1 | - | 5 | 633 | |||
BC070266.1 | - | 5 | 635 | 30418882 |