Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM167B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322248.1 | NP_001309177.1 | 3 | 795 | ||
NM_020141.3 | NP_064526.1 | 3 | 694 | |||
NR_136242.1 | 4 | 823 | ||||
NR_136243.1 | 2 | 602 | ||||
GenBank mRNA | AF164793.1 | - | 3 | 715 | ||
AK075191.1 | - | 3 | 716 | |||
AK310192.1 | - | 2 | 582 | |||
BC008739.2 | - | 3 | 694 | 3346340 | ||
BC065186.1 | - | 3 | 694 | 4642864 | ||
BC066918.1 | - | 3 | 723 | 4826492 | ||
BC067116.1 | - | 3 | 681 | 5532892 | ||
BC171849.1 | - | 3 | 1672 | 9054503 | ||
BG724370.1 | - | 2 | 607 | |||
BX331284.2 | - | 4 | 784 |