Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322854.1 | NP_001309783.1 | 9 | 1398 | ||
NM_001322855.1 | NP_001309784.1 | 10 | 1540 | |||
NM_001322856.1 | NP_001309785.1 | 10 | 1497 | |||
NM_001322857.1 | NP_001309786.1 | 9 | 1318 | |||
NM_001322858.1 | NP_001309787.1 | 10 | 1415 | |||
NM_001322859.1 | NP_001309788.1 | 7 | 1024 | |||
NM_001322860.1 | NP_001309789.1 | 6 | 964 | |||
NM_001322861.1 | NP_001309790.1 | 6 | 674 | |||
NM_001322862.1 | NP_001309791.1 | 10 | 1375 | |||
NM_001322863.1 | NP_001309792.1 | 9 | 1347 | |||
NM_001322864.1 | NP_001309793.1 | 8 | 915 | |||
NM_001322866.1 | NP_001309795.1 | 9 | 1238 | |||
NM_020448.4 | NP_065181.1 | 9 | 1260 | |||
NR_136506.1 | 8 | 1168 | ||||
NR_136508.1 | 8 | 1129 | ||||
XM_011541808.2 | XP_011540110.1 | 8 | 1175 | |||
XM_017001868.1 | XP_016857357.1 | 8 | 910 | |||
XM_017001869.1 | XP_016857358.1 | 9 | 1245 | |||
XM_017001870.1 | XP_016857359.1 | 10 | 1510 | |||
GenBank mRNA | AK312433.1 | - | 9 | 1214 | ||
BC063583.1 | - | 9 | 1190 | 4386115 | ||
BX538110.1 | - | 3 | 288 | |||
BX640883.1 | - | 10 | 1322 | |||
U90918.1 | - | 9 | 1151 |