Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322854.1 | NP_001309783.1 | 11 | 1531 | ||
NM_001322855.1 | NP_001309784.1 | 12 | 1673 | |||
NM_001322856.1 | NP_001309785.1 | 12 | 1630 | |||
NM_001322857.1 | NP_001309786.1 | 11 | 1451 | |||
NM_001322858.1 | NP_001309787.1 | 12 | 1548 | |||
NM_001322859.1 | NP_001309788.1 | 9 | 1157 | |||
NM_001322860.1 | NP_001309789.1 | 8 | 1097 | |||
NM_001322861.1 | NP_001309790.1 | 8 | 807 | |||
NM_001322862.1 | NP_001309791.1 | 12 | 1508 | |||
NM_001322863.1 | NP_001309792.1 | 11 | 1480 | |||
NM_001322864.1 | NP_001309793.1 | 10 | 1048 | |||
NM_001322866.1 | NP_001309795.1 | 11 | 1371 | |||
NM_001322868.1 | NP_001309797.1 | 9 | 1245 | |||
NM_020448.4 | NP_065181.1 | 11 | 1393 | |||
NR_136506.1 | 10 | 1301 | ||||
NR_136507.1 | 5 | 808 | ||||
NR_136508.1 | 10 | 1262 | ||||
XM_011541808.2 | XP_011540110.1 | 10 | 1308 | |||
XM_017001868.1 | XP_016857357.1 | 10 | 1043 | |||
XM_017001869.1 | XP_016857358.1 | 11 | 1378 | |||
XM_017001870.1 | XP_016857359.1 | 12 | 1643 | |||
GenBank mRNA | AK312433.1 | - | 11 | 1347 | ||
BC063583.1 | - | 11 | 1323 | 4386115 | ||
BX538110.1 | - | 5 | 421 | |||
BX640883.1 | - | 12 | 1455 | |||
U90918.1 | - | 11 | 1284 |